missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KISS1R Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15505-1-AP-150UL
This item is not returnable.
View return policy
Description
GCPR54 is a galanin-like G protein coupled receptor that binds metastin, a peptide encoded by the metastasis suppressor gene KISS1. The tissue distribution of the protein suggests that it is involved in the regulation of endocrine function, and this is supported by the finding that it appears to play a role in the onset of puberty. GPR54 has been reported in various human brain regions, placenta, pancreas, and spinal cord, and at lower levels in spleen, peripheral blood leukocytes, testis, lymph node, thymus, small intestine, stomach, lung, and kidney. The receptor is also abundant in tumor tissues. In rat, GPR54 is expressed in liver, intestine, and in the pons, midbrain, thalamus, hypothalamus, hippocampus, amygdala, cortex (including frontal), and striatum. ESTs have been isolated from normal placenta and kidney cancer libraries.Specifications
| KISS1R | |
| Polyclonal | |
| Unconjugated | |
| KISS1R | |
| AXOR12, G protein coupled receptor 54, GPR54, HOT7T175, Hypogonadotropin 1, KiSS 1 receptor, KiSS 1R, KISS1 receptor, KISS1R, Kisspeptins receptor, Metastin receptor | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 84634 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q969F8 | |
| KISS1R | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction