missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Klebsiella Aerogenes Antibody (73/28), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB100-64558CL1
This item is not returnable.
View return policy
Description
Klebsiella Aerogenes Monoclonal antibody specifically detects Klebsiella Aerogenes in Bacteria samples. It is validated for ELISA,Immunocytochemistry/Immunofluorescence
Specifications
| Klebsiella Aerogenes | |
| Monoclonal | |
| CoraFluor 1 | |
| k aerogenes, K. aerogenes | |
| Native Klebsiella aerogenes preparation | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| 73/28 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction