missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF10 Antibody (SR1940), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22109-50ul
This item is not returnable.
View return policy
Description
KLF10 Monoclonal antibody specifically detects KLF10 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| KLF10 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| SR1940 | |
| Western Blot 1:500-1:2000 | |
| EGRA, EGR-alpha, Krueppel-like factor 10, Kruppel-like factor 10, TGFB inducible early growth response, TGFB-inducible early growth response protein 1, TIEG-1, TIEG1TIEG, Transforming growth factor-beta-inducible early growth response protein 1 | |
| A synthesized peptide derived from human KLF10 (Uniprot #: Q13118) | |
| 50 μg | |
| Cell Cycle and Replication | |
| 7071 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction