missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLRG1 Antibody (SR1639), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22188-50ul
This item is not returnable.
View return policy
Description
KLRG1 Monoclonal antibody specifically detects KLRG1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| KLRG1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| SR1639 | |
| Western Blot 1:500-1:2000 | |
| 2F1, CLEC15AMAFA-LIKE, C-type lectin domain family 15 member A, C-type lectin domain family 15, member A, ITIM-containing receptor MAFA-L, killer cell lectin-like receptor subfamily G member 1, killer cell lectin-like receptor subfamily G, member 1, MAFAL, MAFA-L, MAFA-like receptor, MAFAMAFA-2F1, Mast cell function-associated antigen, mast cell function-associated antigen (ITIM-containing), MGC13600 | |
| A synthesized peptide derived from human KLRG1 (Uniprot #: Q96E93) | |
| 50 μg | |
| Immunology, Innate Immunity, Signal Transduction | |
| 10219 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction