missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLRG1 Golden Hamster, PE, Clone: 2F1, enQuireBio™
Golden Hamster Monoclonal Antibody
156.45€ - 367.50€
Specifications
| Antigen | KLRG1 |
|---|---|
| Clone | 2F1 |
| Concentration | 0.5mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15932428
|
EnquireBio
QAB74-PE-25ug |
25 μg |
156.45€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15922428
|
EnquireBio
QAB74-PE-100ug |
100 μg |
367.50€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| KLRG1 | |
| 0.5mg/mL | |
| Monoclonal | |
| Golden Syrian Hamster | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
| 50928 | |
| IgG | |
| Protein A/G Purified |
| 2F1 | |
| Flow Cytometry | |
| PE | |
| RUO | |
| NP_058666 | |
| Klrg1 | |
| Primary | |
| Store at 2-4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title