missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KMT2D Antibody (CL5346) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61416PE
This item is not returnable.
View return policy
Description
KMT2D Monoclonal antibody specifically detects KMT2D in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| KMT2D | |
| Monoclonal | |
| PE | |
| AAD10, ALRALL1-related protein, CAGL114, EC 2.1.1.43, histone-lysine N-methyltransferase MLL2, KMT2D, Lysine N-methyltransferase 2B, MLL4, myeloid/lymphoid or mixed-lineage leukemia 2, Myeloid/lymphoid or mixed-lineage leukemia protein 2, TNRC21, trinucleotide repeat containing 21 | |
| This antibody was developed using a recombinant protein derived from O14686, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunocytochemistry/Immunofluorescence | |
| CL5346 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 8085 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction