Learn More
Koflo Tube Mixer
Product Code. 11774119 Shop All Cole-Parmer Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Length (Metric):
178 mm
190 mm
292 mm
302 mm
missing translation for 'unitSize'
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
11774119 178 mm Each
11764119 190 mm Each
11794119 292 mm Each
11730029 302 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 missing translation for 'options'
Product Code. 11774119 missing translation for 'mfr' Koflo missing translation for 'supplierNo' 0466956
  • ONLINE EXCLUSIVE PRICE
    247.00€
    216.54€ / Pack of 1
    Save 30.46€
    12% Off
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
Use Tube Mixer to mix fluids as you pump them through a pipe line.
  • Able to mix light viscosity to high viscosity components.
  • Accept standard tube fittings.
  • These mixers consist of a series of fixed right- and left-hand elements.
  • Tube and mixing elements are made of 316 stainless steel—ideal for mixing high-viscosity fluids such as epoxies.
  • Mixers have plain ends and accept standard tube fittings.
Specifications
Diameter (English) 0.24 in.
Diameter (Metric) 6 mm
Length (English) 7 in.
Length (Metric) 178 mm
Material Stainless Steel (316)
Type Tube Mixer
For Use With (Application) Mixing high-viscosity fluids