Learn More
DWK Life Sciences Kimble™ Kontes™ Chromatography Column ETFE Nut for 1/16 in. Tubing
Product Code. 11752344
Change view
Click to view available options
For Use With:
Flex-Column
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With Unit Size
11752344 Flex-Column Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11752344 Supplier DWK Life Sciences Supplier No. 4208210116
  • ONLINE EXCLUSIVE PRICE
    6.99€
    5.47€ / Pack of 1
    Save 1.52€
    21% Off
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Tubing nuts and ferrules are used to make connections between the tubing and the end fittings of CHROMAFLEX™, FLEX-COLUMN™ and DISPOSAFLEX™ columns.
  • ETFE Flangeless Nut
  • 1/4-28 Thread, for 1/16 in. Tubing
  • Use with the 420822-Series Flangeless Ferrules
  • Case of 1
Specifications
Type Flangeless Nut
Diameter (English) Outer 0.0625 in.
Diameter (Metric) Outer 0.158 cm
Thread Size 0.25-28
For Use With Flex-Column
Material ETFE