Learn More
DWK LIFE SCIENCES™ KIMBLE™ KONTES™ NMR Polyethylene Sample Tube Pressure Caps for 3, 5 and 10mm Tubes
Description
- Tight-fitting polyethylene construction stays in place even with a build-up of internal vaporization pressures
- Available for 3mm, 5mm and 10mm NMR sample tubes
- Available in a variety of colors for easy sample identification and organization
- Assortment packs include a variety of colors in each pack
- Kimble™ Kontes™ brand quality from DWK Life Sciences
Specifications
Specifications
| Closure Size | 5 mm |
| Product Type | Pressure Cap |
| Autoclavable | Not Autoclavable |
| Material | Polyethylene |
| Color | Red |
| For Use With (Application) | Provides tight-fitting and stays in place even with a build-up of internal vaporization pressures |
| Sterility | Non-sterile |
| For Use With (Equipment) | 5 mm NMR Sample Tube |
| Disposable | Reusable |