Learn More
KPNA2 Mouse anti-Human, Unconjugated, Clone: 2G9C1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66870-1-IG-150UL
Description
The KPNA2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human KPNA2. This antibody recognizes human antigen. The KPNA2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| KPNA2 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, ELISA | |
| 2G9C1 | |
| Unconjugated | |
| 150 μL | |
| 3838 | |
| IgG1 |