Learn More
KPNA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10819-1-AP-20UL
This item is not returnable.
View return policy
Description
The import of proteins into the nucleus is a process that involves at least 2 steps. The first is an energy-independent docking of the protein to the nuclear envelope and the second is an energy-dependent translocation through the nuclear pore complex. Imported proteins require a nuclear localization sequence which generally consists of a short region of basic amino acids or 2 such regions spaced about 10 amino acids apart. Proteins involved in the first step of nuclear import have been identified in different systems. These include the Xenopus protein importin and its yeast homolog, SRP1, which bind to the NLS. KPNA2 protein interacts with the NLSs of DNA helicase Q1 and SV40 T antigen and may be involved in the nuclear transport of proteins. KPNA2 also may play a role in VJ recombination.Specifications
| KPNA2 | |
| Polyclonal | |
| Unconjugated | |
| KPNA2 | |
| Importin subunit alpha 2, IPOA1, Karyopherin subunit alpha 2, KPNA2, QIP2, RAG cohort protein 1, RCH1, SRP1, SRP1 alpha, SRP1alpha | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16647, 3838, 85245 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P52292, P52293 | |
| KPNA2 | |
| KPNA2 Fusion Protein Ag1153 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |