Learn More
KRT35 Monoclonal Antibody (OTI1A11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811820S
Description
KRT35 Monoclonal antibody specifically detects KRT35 in Human samples. It is validated for Western BlotSpecifications
| KRT35 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q6ZMV9 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 221458 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI1A11 | |
| Unconjugated | |
| KIF6 | |
| AI662400; Ha5; Ha-5; hair keratin; hair keratin acidic 5; hair keratin, type I Ha5; hard keratin, type I, 5; HHA5; HHa5 hair keratin type I intermediate filament; HKA5; K35; Ka30; keratin 35; keratin 35, type I; keratin complex 1, acidic, gene 2; keratin complex 1, acidic, gene 24; keratin complex 1, gene 24; keratin, hair, acidic, 5; keratin, type I cuticular Ha5; Keratin-35; KRT hHa5; Krt1-2; Krt1-24; KRT35; KRTHA5; type I hair keratin KA30; Unknown (protein for MGC:139866); wool (hair) keratin intermediate filament protein | |
| Human recombinant protein fragment corresponding to amino acids 1-350 of human KIF6 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.