missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KRT78 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558902
This item is not returnable.
View return policy
Description
Immunogen sequence: VGGGLGSTCG LGSGKGSPGS CCTSIVTGGS NIILGSGKDP VLDSCSVSGS S Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 52%.
This gene is a member of the type II keratin gene family and encodes a protein with an intermediate filament domain. Keratins are the major structural proteins in epithelial cells, forming a cytoplasmic network of 10 to 12 nm wide intermediate filaments and creating a scaffold that gives cells the ability to withstand mechanical and non-mechanical stresses. The genes of the type II keratin family are located as a gene cluster at 12p13.13. Four pseudogenes of this gene family have been identified.
Specifications
| KRT78 | |
| Polyclonal | |
| Unconjugated | |
| KRT78 | |
| CK-78; Cytokeratin-78; K5B; K78; Kb40; keratin 5b; keratin 78; keratin 78, type II; keratin, type II cytoskeletal 78; Keratin-5b; Keratin-78; KRT78; Type-II keratin Kb40 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 196374 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N1N4 | |
| KRT78 | |
| Recombinant protein corresponding to Human KRT78. Recombinant protein control fragment (Product #RP-97817). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction