missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KSHV ORF62 Monoclonal Antibody (5B7B6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA514770
This item is not returnable.
View return policy
Description
MA5-14770 targets KSHV ORF62 in indirect ELISA, IHC and WB applications and shows reactivity with Virus samples. The MA5-14770 immunogen is purified recombinant fragment of human KSHV ORF62 expressed in E. Coli.
Kaposi's sarcoma-associated herpesvirus (KSHV) belongs to the gamma-(2)-herpesvirus subfamily and has been closely linked to the Kaposi's sarcoma, primary effusion lymphoma (PEL) and multicentric Castleman's disease. The genome of KSHV is 165-170 kb and contains at least 88 open reading frames. At least five major proteins are likely to be involved in the assembly of the HHV-8 capsid, including a protease (encoded by ORF17), the major capsid protein (encoded by ORF25), and three other smaller capsid proteins (encoded by ORF62, ORF26, and ORF65). Previous structural studies have shown that the HSV-1 triplex is a monomer of VP19c and a dimer of VP23 and that the HCMV triplex is similarly composed of a monomer and a dimer. By analogy, the KSHV triplexes are likely also composed of a monomer of the ORF62 protein and a dimer of the ORF26 protein, which are the respective homologs of VP19c and VP23.
Specifications
| KSHV ORF62 | |
| Monoclonal | |
| Conc. Not Determined | |
| ascites with 0.03% sodium azide | |
| Mouse | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 5B7B6 | |
| Unconjugated | |
| Kaposi's Sarcoma Virus; Kaposi's sarcoma-associated herpesvirus | |
| Purified recombinant fragment of human KSHV ORF62 expressed in E. Coli. | |
| RUO | |
| Virus | |
| Antibody | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction