Learn More
Ku80/XRCC5 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-508PCP
This item is not returnable.
View return policy
Description
Ku80/XRCC5 Polyclonal antibody specifically detects Ku80/XRCC5 in Human, Mouse, Rat, Hamster samples. It is validated for Western Blot, Immunoprecipitation, ChIP assay (ChIP)
Specifications
| ChIP sequencing (ChIP-seq) | |
| PerCP | |
| A synthetic peptide corresponding to residues 419-440 LVYVQLPFMEDLRQYMFSSLKN] of the 80 kDa Ku80 protein. | |
| 7520 |
| Polyclonal | |
| PBS | |
| RUO | |
| IgG |