Learn More
Ku80/XRCC5 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB100-508PCP
Product Code. 18543193
  • 542.00€ / 0.10mL
Description
Ku80/XRCC5 Polyclonal antibody specifically detects Ku80/XRCC5 in Human, Mouse, Rat, Hamster samples. It is validated for Western Blot, Immunoprecipitation, ChIP assay (ChIP)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ChIP sequencing (ChIP-seq)
PerCP
A synthetic peptide corresponding to residues 419-440 LVYVQLPFMEDLRQYMFSSLKN] of the 80 kDa Ku80 protein.
7520