missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ L-651,582

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Orally active Ca2+ channel blocker; inhibits K+- and carbachol-stimulated Ca2+ influx (IC50 values are 500 and 935 nM respectively). Exhibits antiproliferative, antiangiogenic and antimetastatic activity in vivo and displays selectivity towards numerous mismatch repair-deficient tumor cell lines in vitro.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 424.67 |
| Purity | >98% |
| Molecular Weight (g/mol) | 424.67 |
| Chemical Name or Material | L-651, 582 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction