missing translation for 'onlineSavingsMsg'
Learn More

L-(-)-Glucose, anhydrous, 98%, Thermo Scientific Chemicals

Product Code. 11458107
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11458107

Brand: Thermo Scientific Alfa Aesar A17496.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

L-(-)-Glucose is an enantiomer of the more common D-glucose a naturally occurring carbohydrate used in an abundance of cellular processes. L-Glucose is a synthetic sugar used in the formation of L-Glucose Pen taacetate, a potential therapeutic agent regarding type II diabetes. In addition, L-glucose can be used as a colon cleansing agent for before a colonoscopy procedure.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-(-)-Glucose is an enantiomer of the more common D-glucose a naturally occurring carbohydrate used in an abundance of cellular processes. L-Glucose is a synthetic sugar used in the formation of L-Glucose Pen taacetate, a potential therapeutic agent regarding type II diabetes. In addition, L-glucose can be used as a colon cleansing agent for before a colonoscopy procedure.

Solubility
Soluble in water, alcohol.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 921-60-8
Molecular Formula C6H12O6
MDL Number MFCD00148913
PubChem CID 10954115
ChEBI CHEBI:37626

Specifications

Melting Point 148°C to 157°C
Quantity 5 g
Beilstein 1724626
Synonym l-glucose, l---glucose, levoglucose, l--glucose, 2 s,3 r,4 s,5 s-2,3,4,5,6-pentahydroxyhexanal, aldehydo-l-glucose, aldehydo-l-gluco-hexose, d-glucose-13c6,d7, levoglucose usan, glucose, l-isomer
Solubility Information Soluble aq. soln.,alcohol.
InChI Key WQZGKKKJIJFFOK-UHFFFAOYNA-N
Optical Rotation -53°C=10 aq. soln., 24 h)
SMILES OCC1OC(O)C(O)C(O)C1O
IUPAC Name 6-(hydroxymethyl)oxane-2,3,4,5-tetrol
Molecular Weight (g/mol) 180.16
Formula Weight 180.16
Percent Purity 98%
Chemical Name or Material L-(-)-Glucose, anhydrous
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.