missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Glutamic-2,3,3,4,4-d5 Acid, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | L-Glutamic Acid-d5 |
| CAS | 2784-50-1 |
| Molecular Formula | C5 2H5 H4 N O4 |
| Quantity | 0.1 g |
| InChI Formula | InChI=1 S/C5H9NO4/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)/t3-/m0/s1/i1D2,2D2,3 D |
| SMILES | [2 H]C([2 H])(C(=O)O)C([2 H])([2 H])[C@]([2 H])(N)C(=O)O |
| IUPAC Name | (2 S)-2-amino-2,3,3,4,4-pentadeuteriopentanedioic acid |
| Molecular Weight (g/mol) | 152.16 |
| Formula Weight | 152.0845 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction