missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic Acid 5-Methyl Ester, TRC

Product Code. 30396295
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396295 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30396295 Supplier Toronto Research Chemicals Supplier No. TRCG59702010G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Glutamic Acid 5-Methyl Ester
CAS 1499-55-4
Molecular Formula C6 H11 N O4
Quantity 10 g
Synonym (S)-2-Aminopentanedioic Acid 5-Methyl Ester, 4(S)-Carboxy-4-aminobutanoic Acid Methyl Ester, 5-Methyl L-Glutamate, Glutamic Acid 5-Methyl Ester, L-Glutamic Acid gamma-Methyl Ester, NSC 12959, gamma-Methyl (S)-Glutamate, gamma-Methyl L-Glutamate, gamma-Methyl L-Glutamate
InChI Formula InChI=1S/C6H11NO4/c1-11-5(8)3-2-4(7)6(9)10/h4H,2-3,7H2,1H3,(H,9,10)/t4-/m0/s1
SMILES COC(=O)CC[C@H](N)C(=O)O
IUPAC Name (2S)-2-amino-5-methoxy-5-oxopentanoic acid
Molecular Weight (g/mol) 161.156
Formula Weight 161.069
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.