missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamine, USP Grade, MP Biomedicals™

Product Code. p-7087643 Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 11408382

Brand: MP Biomedicals 0219960890

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 56-85-9
Molecular Formula C5H10N2O3
Molecular Weight (g/mol) 146.15
MDL Number MFCD00008044
InChI Key ZDXPYRJPNDTMRX-VKHMYHEASA-N
Synonym (S)-2,5-Diamino-5-oxopentanoic acid; L-Glutamic acid 5-amide
IUPAC Name (2S)-2-amino-4-carbamoylbutanoic acid
SMILES N[C@@H](CCC(N)=O)C(O)=O

Specifications

Melting Point 185.5°C
pH 5 to 6 (at 14.6 g/L, 25°C)
Color White
Beilstein 1723797
Density 1.364 g/cm3
Quantity 500 g
Optical Rotation (40 mg/mL aq. soln. (at 20°C) +6.3° to +7.3°)
Grade USP
Physical Form Powder
Chemical Name or Material L-Glutamine, USP

For research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.