missing translation for 'onlineSavingsMsg'
Learn More

L-Isoleucinol, 97%

Product Code. 15457038
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15457038 1 g 1g
15467038 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15457038 Supplier Thermo Scientific Alfa Aesar Supplier No. H60556.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 24629-25-2 | C6H16NO | 118.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24629-25-2
Molecular Formula C6H16NO
Molecular Weight (g/mol) 118.20
MDL Number MFCD00004731
InChI Key VTQHAQXFSHDMHT-NTSWFWBYSA-O
Synonym l-isoleucinol, 2s,3s-2-amino-3-methylpentan-1-ol, s-+-isoleucinol, h-isoleucinol, l-+-isoleucinol, isoleucinol, 2s,3s-2-amino-3-methyl-pentan-1-ol, h-ile-ol, 1-pentanol, 2-amino-3-methyl-, 2s,3s
PubChem CID 2724027
IUPAC Name (2S,3S)-2-amino-3-methylpentan-1-ol
SMILES CC[C@H](C)[C@H]([NH3+])CO

Specifications

Refractive Index 1.4589
Boiling Point 97°C
Melting Point 36°C to 39°C
Quantity 1 g
Flash Point 100°C (212°F)
Linear Formula C2H5CH(CH3)CH(NH2)CH2OH
Formula Weight 117.19
Percent Purity 97%
Chemical Name or Material L-Isoleucinol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.