missing translation for 'onlineSavingsMsg'
Learn More

L-Norvalyl diphenylborinate, 95%, Thermo Scientific™

Product Code. 15451588
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15451588 250 mg 250mg
15441588 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15451588 Supplier Thermo Scientific Alfa Aesar Supplier No. H55368.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17093-22-0
Molecular Formula C17H20BNO2
Molecular Weight (g/mol) 281.162
MDL Number MFCD22125141
InChI Key XAVUFSLMIUPEFM-INIZCTEOSA-N
Synonym l-norvalyl diphenylborinate, diphenylboranyl 2s-2-aminopentanoate
PubChem CID 99738133
IUPAC Name diphenylboranyl (2S)-2-aminopentanoate
SMILES B(C1=CC=CC=C1)(C2=CC=CC=C2)OC(=O)C(CCC)N

Specifications

Quantity 250 mg
Formula Weight 281.16
Percent Purity 95%
Chemical Name or Material L-Norvalyl diphenylborinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.