missing translation for 'onlineSavingsMsg'
Learn More

L-Prolinamide, TRC

Product Code. 17988024
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17988024 5 g 5g
17938994 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17988024 Supplier Toronto Research Chemicals Supplier No. TRCP7559755G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7531-52-4
Molecular Formula C5 H10 N2 O
Molecular Weight (g/mol) 114.146
Synonym 2-Pyrrolidinecarboxamide, (2S)-, 2-Pyrrolidinecarboxamide, (S)-, Prolinamide, L- (8CI), (2S)-2-Pyrrolidinecarboxamide, (-)-Prolinamide, (2S)-2-Carbamoylpyrrolidine, (2S)-Pyrrolidine-2-carboxylic acid amide, (S)-2-(Aminocarbonyl)pyrrolidine, (S)-Prolinamide, (S)-Proline amide, (S)-Pyrrolidine-2-carboxamide, (S)-Pyrrolidine-2-carboxylic acid amide, L-Prolinamide, L-Proline amide
IUPAC Name (2S)-pyrrolidine-2-carboxamide
SMILES NC(=O)[C@@H]1CCCN1

Specifications

InChI Formula InChI=1S/C5H10N2O/c6-5(8)4-2-1-3-7-4/h4,7H,1-3H2,(H2,6,8)/t4-/m0/s1
Quantity 5 g
Formula Weight 114.0793
Shipping Condition Room Temperature
Chemical Name or Material L-Prolinamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.