missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Selectin/CD62L Antibody (DREG-56), mFluor Violet 500 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81084MFV500
492.15 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
L-Selectin/CD62L Monoclonal antibody specifically detects L-Selectin/CD62L in Human, Bovine, Chimpanzee samples. It is validated for Flow Cytometry
Specifications
| L-Selectin/CD62L | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Bovine, Chimpanzee | |
| Purified |
| Flow Cytometry | |
| DREG-56 | |
| Flow Cytometry | |
| CD62L, CD62L antigen, gp90-MEL, hLHRc, LAM-1, LAM1LECAM1, Leu-8, LEU8, Leukocyte adhesion molecule 1, Leukocyte surface antigen Leu-8, Leukocyte-endothelial cell adhesion molecule 1, LNHRTQ1, LSEL, L-selectin, Lyam-1, LYAM1CD62 antigen-like family member L, Lymph node homing receptor, lymphocyte adhesion molecule 1, pln homing receptor, PLNHR, selectin L | |
| This antibody was produced by immunizing mice with the concentrated supernatant generated after centrifugation of human peripheral blood leukocytes. | |
| 0.1 mL | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cardiovascular Biology, Glycobiology, Immunology, Lipid and Metabolism, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cells | |
| 6402 | |
| Store at 4C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction