missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-selectin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26477-1-AP-150UL
This item is not returnable.
View return policy
Description
The human CD62L is a 74-95 kD glycoprotein member of the selectin family of adhesion receptors. L-Selectin is comprised of an aminoterminal C-type lectin binding domain, an epidermal growth factor-like domain, two short consensus repeat (SCR) sequences homologous to those found in complement binding proteins, a short spacer region, a transmembrane region and a short cytoplasmic region. Human CD62L (L-Selectin) is constitutively expressed on all classes of leukocytes including lymphocytes (except a substantial population of memory T cells), monocytes and polymorphonuclear cells.Specifications
| L-selectin | |
| Polyclonal | |
| Unconjugated | |
| SELL | |
| CD62L, gp90 MEL, hLHRc, LAM 1, LAM1, LECAM1, Leu 8, Leukocyte adhesion molecule 1, LNHR, LSEL, L-selectin, Lyam 1, LYAM1, Lymph node homing receptor, PLNHR, selectin L, SELL, TQ1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6402 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P14151 | |
| SELL | |
| L-selectin Fusion Protein Ag24063 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction