Learn More
Copan Diagnostics L-Shaped Spreaders
Product Code. 11726548
Change view
Click to view available options
Quantity:
1/Peel Pk., 500 Spreaders/Box
5/Peel Pk., 1000 Spreaders/Box
Unit Size:
Pack of 1000
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11726548 1/Peel Pk., 500 Spreaders/Box Pack of 500
11736548 5/Peel Pk., 1000 Spreaders/Box Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11726548 Supplier Copan Diagnostics Supplier No. 174CS01
  • ONLINE EXCLUSIVE PRICE
    243.00€
    113.77€ / Pack of 500
    Save 129.23€
    53% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
L-shaped spreaders
L-Shaped Plastic Spreaders
  • For spreading liquid samples
  • Foot of the spreader has a completely smooth, rounded surface, free of rough edges and imperfections in the plastic
  • Enables even spreading of liquid samples across the surface of agar plates without gouging or cutting the medium
  • Available in two sterile peel pack sizes, five or ten spreaders per pack
Specifications
For Use With (Equipment) Culture Plates
Quantity 1/Peel Pk., 500 Spreaders/Box
Shape L-Shape
Material Plastic
Product Type Cell Spreader
Sterility Sterile
For Use With (Application) Designed for spreading liquid samples, enables even spreading of liquid samples across the surface of agar plates without gouging or cutting the medium