Learn More
Copan Diagnostics L-Shaped Spreaders
L-shaped spreaders
195.00€ - 225.00€
Specifications
Material | Plastic |
---|---|
Sterility | Sterile |
For Use With (Equipment) | Culture Plates |
Shape | L-Shape |
For Use With (Application) | Designed for spreading liquid samples, enables even spreading of liquid samples across the surface of agar plates without gouging or cutting the medium |
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
11726548
|
Copan Diagnostics
174CS01 |
1/peel pack; 500 spreaders/box |
195.00€
Pack of 500 |
In Stock
Log in to see stock. |
|||||
11736548
|
Copan Diagnostics
174CS05 |
5/peel pack; 1000 spreaders/box |
225.00€
Pack of 1000 |
In Stock
Log in to see stock. |
|||||
Description
L-shaped spreaders
L-Shaped Plastic Spreaders
- For spreading liquid samples
- Foot of the spreader has a completely smooth, rounded surface, free of rough edges and imperfections in the plastic
- Enables even spreading of liquid samples across the surface of agar plates without gouging or cutting the medium
- Available in two sterile peel pack sizes, five or ten spreaders per pack
Specifications
Plastic | |
Culture Plates | |
Designed for spreading liquid samples, enables even spreading of liquid samples across the surface of agar plates without gouging or cutting the medium |
Sterile | |
L-Shape |