Learn More
L-Threoninol, TRC
Product Code. 30449301
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30449301 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30449301 Supplier Toronto Research Chemicals Supplier No. TRCT405578500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Ajouter au panier
Ajouter au panier
missing translation for 'validMainframeMsg'
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Spécification
Chemical Name or Material L-Threoninol
CAS 3228-51-1
Molecular Formula C4H11NO2
Quantity 500 mg
InChI Formula InChI=1S/C4H11NO2/c1-3(7)4(5)2-6/h3-4,6-7H,2,5H2,1H3/t3-,4-/m1/s1
SMILES C[C@@H](O)[C@H](N)CO
IUPAC Name (2R,3R)-2-aminobutane-1,3-diol
Molecular Weight (g/mol) 105.14
Formula Weight 105.079