missing translation for 'onlineSavingsMsg'
Learn More

L-Thyroxine, 98%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J62606.06

Quantity 5g

 View more versions of this product

Product Code. 15474939

  • 320.00€ / 5g
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

51-48-9
776.87
XUIIKFGFIJCVMT-UHFFFAOYNA-N
5819
2-amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propanoic acid
C15H11I4NO4
MFCD00002595
l-thyroxine, levothyroxine, thyroxine, thyroxin, synthroid, tetraiodothyronine, thyrax, levothyroxin, thyratabs, thyreoideum
CHEBI:18332
NC(CC1=CC(I)=C(OC2=CC(I)=C(O)C(I)=C2)C(I)=C1)C(O)=O

Specifications

51-48-9
C15H11I4NO4
5g
Light sensitive
l-thyroxine, levothyroxine, thyroxine, thyroxin, synthroid, tetraiodothyronine, thyrax, levothyroxin, thyratabs, thyreoideum
XUIIKFGFIJCVMT-UHFFFAOYNA-N
2-amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propanoic acid
5819
776.87
L-Thyroxine
Odorless
MFCD00002595
2228515
14,9415
Dissolves in 4M ammonium hydroxide in Methanol at 50mg/ml
NC(CC1=CC(I)=C(OC2=CC(I)=C(O)C(I)=C2)C(I)=C1)C(O)=O
776.87
CHEBI:18332
98%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.