Learn More
L-Thyroxine, 98%, Thermo Scientific™
85.65€ - 320.00€
Chemical Identifiers
| CAS | 51-48-9 |
|---|---|
| Molecular Formula | C15H11I4NO4 |
| Molecular Weight (g/mol) | 776.87 |
| MDL Number | MFCD00002595 |
| InChI Key | XUIIKFGFIJCVMT-UHFFFAOYNA-N |
| Synonym | l-thyroxine, levothyroxine, thyroxine, thyroxin, synthroid, tetraiodothyronine, thyrax, levothyroxin, thyratabs, thyreoideum |
| PubChem CID | 5819 |
| ChEBI | CHEBI:18332 |
| IUPAC Name | 2-amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propanoic acid |
| SMILES | NC(CC1=CC(I)=C(OC2=CC(I)=C(O)C(I)=C2)C(I)=C1)C(O)=O |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15464939
|
Thermo Scientific Alfa Aesar
J62606.03 |
1g |
94.80€ 85.65€ / 1g Save 9.15€ 9% Off |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
15474939
|
Thermo Scientific Alfa Aesar
J62606.06 |
5g |
320.00€
5g |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 51-48-9 | |
| 776.87 | |
| XUIIKFGFIJCVMT-UHFFFAOYNA-N | |
| 5819 | |
| 2-amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propanoic acid |
| C15H11I4NO4 | |
| MFCD00002595 | |
| l-thyroxine, levothyroxine, thyroxine, thyroxin, synthroid, tetraiodothyronine, thyrax, levothyroxin, thyratabs, thyreoideum | |
| CHEBI:18332 | |
| NC(CC1=CC(I)=C(OC2=CC(I)=C(O)C(I)=C2)C(I)=C1)C(O)=O |
Specifications
| 51-48-9 | |
| C15H11I4NO4 | |
| 2228515 | |
| 14,9415 | |
| Dissolves in 4M ammonium hydroxide in Methanol at 50mg/ml | |
| NC(CC1=CC(I)=C(OC2=CC(I)=C(O)C(I)=C2)C(I)=C1)C(O)=O | |
| 776.87 | |
| CHEBI:18332 | |
| 98% |
| Odorless | |
| MFCD00002595 | |
| Light sensitive | |
| l-thyroxine, levothyroxine, thyroxine, thyroxin, synthroid, tetraiodothyronine, thyrax, levothyroxin, thyratabs, thyreoideum | |
| XUIIKFGFIJCVMT-UHFFFAOYNA-N | |
| 2-amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propanoic acid | |
| 5819 | |
| 776.87 | |
| L-Thyroxine |
Safety and Handling
P264-P270-P301+P312-P330-P501c
H302
missing translation for 'einecsNumber' : 200-101-1
missing translation for 'rtecsNumber' : YP2833500
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.