missing translation for 'onlineSavingsMsg'
Learn More

L-Thyroxine, MP Biomedicals™

Product Code. 11498301 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11498301 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Product Code. 11498301 Supplier MP Biomedicals Supplier No. 0215214505
  • ONLINE EXCLUSIVE PRICE
    519.00€
    493.50€ / 5g
    Save 25.50€
    4% Off
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

Specifications

CAS 51-48-9
Color White
Molecular Formula C15H11I4NO4
MDL Number MFCD00002595
Quantity 5 g
Synonym l-thyroxine, levothyroxine, thyroxine, thyroxin, synthroid, tetraiodothyronine, thyrax, levothyroxin, thyratabs, thyreoideum
InChI Key XUIIKFGFIJCVMT-UHFFFAOYNA-N
SMILES NC(CC1=CC(I)=C(OC2=CC(I)=C(O)C(I)=C2)C(I)=C1)C(O)=O
IUPAC Name 2-amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propanoic acid
Molecular Weight (g/mol) 776.87
PubChem CID 5819
ChEBI CHEBI:18332
Formula Weight 776.9
Physical Form Solid
Chemical Name or Material L-Thyroxine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.