missing translation for 'onlineSavingsMsg'
Learn More

L-Valine-2-d1, CDN

Product Code. 30312200
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30312200

Brand: CDN CDND54810.25G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material L-Valine-2-d1
CAS 77257-03-5
Molecular Formula C5 D H10 N O2
Quantity 0.25 g
Synonym L-Valine-2-d (9 CI, ACI), L-(α-2 H)Valine, (2 S)-2-Amino-3-methylbutanoic-2-d acid, L-Valine-2-D
InChI Formula InChI=1 S/C5H11NO2/c1-3(2)4(6)5(7)8/h3-4 H,6H2,1-2H3,(H,7,8)/t4-/m0/s1/i4D
SMILES [2 H][C@](N)(C(C)C)C(=O)O
IUPAC Name (2 S)-2-amino-2-deuterio-3-methylbutanoic acid
Molecular Weight (g/mol) 118.153
Formula Weight 118.085 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity
Visa mer Visa mindre
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado