missing translation for 'onlineSavingsMsg'
Learn More

L-xylo-2-Hexulosonic Acid Methyl Ester, TRC

Product Code. 30457085
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457085 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457085 Supplier Toronto Research Chemicals Supplier No. TRCX750315500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl L-Sorbosonate
CAS 3031-98-9
Molecular Formula C7 H12 O7
Quantity 500 mg
Synonym L-xylo-2-Hexulosonic acid','methyl ester, Gulonic acid','2-keto-','methyl ester','L- (6CI), L-xylo-Hexulosonic acid','methyl ester (7CI,8CI), 2-keto-L-Gulonic acid methyl ester, Methyl 2-keto-L-gulonate, Methyl L-xylo-hexulosonate, NSC 163924, Methyl L-Sorbosonate, Methyl L-xylo-Hex-2-ulosonate, Ascorbic Acid Imp. D (EP)
InChI Formula InChI=1S/C7H12O7/c1-14-7(13)6(12)5(11)4(10)3(9)2-8/h3-5,8-11H,2H2,1H3/t3-,4+,5-/m0/s1
SMILES COC(=O)C(=O)[C@@H](O)[C@H](O)[C@@H](O)CO
IUPAC Name methyl (3S,4R,5S)-3,4,5,6-tetrahydroxy-2-oxohexanoate
Molecular Weight (g/mol) 208.166
Formula Weight 208.0583
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.