missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L1CAM Antibody (CL12988) [DyLight 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18553V
This item is not returnable.
View return policy
Description
L1CAM Monoclonal antibody specifically detects L1CAM in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| L1CAM | |
| Monoclonal | |
| DyLight 405 | |
| antigen identified by monoclonal R1, CAML1N-CAML1, CD171, CD171 antigen, HSAS, HSAS1, L1 cell adhesion molecule, MASA, MIC5, N-CAM-L1, NCAM-L1, neural cell adhesion molecule L1, S10, SPG1 | |
| This antibody was developed using a synthetic peptide derived from P32004, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cellular Markers, Glia Markers, Neuroscience, Phospho Specific | |
| 3897 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL12988 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction