Learn More
Berthold Technologies Lab Jack With Table 200 x 200 mm
Product Code. 31777305 Shop All Berthold Technologies Products
Change view
Click to view available options
For Use With (Equipment):
Imaging System
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
31777305 Imaging System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31777305 Supplier Berthold Technologies Supplier No. 64469
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Used in the NightSHADE imaging system to bring the samples closer to the camera.
  • The camera position in the Berthold NightSHADE in vivo imaging system is fixed
  • The lab jack raises the sample closer to the camera
  • Supports imaging of small samples
  • Allows imaging of specific areas of larger samples
Specifications
Type Support Stand
For Use With (Equipment) Imaging System
Depth (Metric) 200 mm
Length (Metric) 200 mm