Learn More
Epredia™ Lab Vision™ Biotin Ab-2 Mouse Monoclonal Antibody, 200μg/mL with BSA and azide
Detect both the free and protein-conjugated forms of biotin with Epredia™ Biotin Ab-2, Mouse Monoclonal Antibody.
Brand: Epredia™ MS-1048-P1
Additional Details : Weight : 0.00190kg
Description
Ab-1 has a very high affinity for biotin and is excellent for use in various amplification techniques. It recognizes both the free and protein-conjugated (either soluble or cell bound) form of biotin. In some applications, localization of biotinylated probes with avidin produces unacceptably high background staining. Anti-biotin antibody may be substituted to decrease this noise.Host Species: Mouse
Clone: BTN.4
Isotype: IgG1/κ
Species Reactivity: All species.
Immunogen: Biotin conjugated to BSA
Recommended for:
- Western Blotting
- Confocal Microscopy
- Flow Cytometry
- Image Analysis
- In situ Nucleic Acid Hybridization
Note:
USA: RUO; Int'l: RUO
Specifications
Biotin Ab-2 | |
Monoclonal | |
200μg/mL | |
Mouse | |
500 μL | |
Epitope Tags | |
All species | |
IgG2a κ |
Western Blot | |
BTN.4 | |
Unconjugated | |
Biotin conjugated to BSA | |
RUO | |
Primary | |
Concentrated |