missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-myc Ab-2 Mouse Monoclonal Antibody, Epredia™
Epredia™ c-myc Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.
133.62€ - 576.30€
Specifications
Antigen | c-myc Ab-2 |
---|---|
Clone | 9E10.3 |
Applications | Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Host Species | Mouse |
Product Code | Brand | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
12629766
|
Epredia
MS-139-P1 |
Concentrated | Unconjugated | 500 μL |
391.00€
500µL |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12689756
|
Epredia
MS-139-B1 |
Concentrated | Biotin | 500 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
12609766
|
Epredia
MS-139-P0 |
Concentrated | Unconjugated | 100 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
12619766
|
Epredia
MS-139-P1ABX |
Concentrated | Unconjugated | 100 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
12639766
|
Epredia
MS-139-PABX |
Concentrated | Unconjugated | 200 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
12679756
|
Epredia
MS-139-B0 |
Concentrated | Biotin | 100 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
12699756
|
Epredia
MS-139-P |
Concentrated | Unconjugated | 1 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
12679766
|
Epredia
MS-139-R7 |
Prediluted | Unconjugated | 7 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
Description
c-myc is involved in the control of cell proliferation and differentiation and is amplified and/or overexpressed in a variety of tumors.Host Species: Mouse
Clone: 9E10.3
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with mouse, rat and chicken. Others not tested.
Epitope: aa 410-419
Immunogen: A synthetic peptide, aa 408-439, (AEEQKLISEEDLLRKRREQLKHKLEQLRNSCA) from C-terminus of human c-myc
Molecular Weight: 64-67 kDa
Positive Control: Raji cells, Breast CA
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Flow Cytometry
- Electron Microscopy
- Affinity Purification, Immunoprecipitation and Western Blotting of fusion proteins labeled with EQKLISEEDL tag
Note:
USA: RUO; Int'l: RUO
Specifications
c-myc Ab-2 | |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Mouse | |
Transcription Regulation | |
A synthetic peptide, aa 408-439, (AEEQKLISEEDLLRKRREQLKHKLEQLRNSCA) from C-terminus of human c-myc | |
Primary |
9E10.3 | |
Monoclonal | |
RUO | |
Human | |
IgG2a κ |