missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Calponin-1 Rabbit Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry procedures with Epredia™ Calponin-1, Rabbit Monoclonal Antibody.

Brand:  Epredia™ RM-2102-RQ

 View more versions of this product

Product Code. 12633607

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

This item is not returnable. View return policy

Description

Description

Calponin, a thin filament associated protein, is implicated in the regulation and modulation of smooth muscle contraction. It is capable of binding to actin, calmodulin, troponin C and tropomyosin. Calponin is expressed in smooth muscle and tissues containing significant amounts of smooth muscle. Two isoforms of calponin exist whose molecular weights are 34 kDa and 29 kDa. Expression of the 29 kDa form is primarily restricted to muscle of the urogenital tract. The expression of calponin has also been demonstrated in myoepithelial cells from benign and malignant breast lesions. It stains smooth muscle, myoepithelial cells, myofibroblasts, keratinocytes and nerve fibers. It identifies myoepithelial cells in breast lesions, and helps differentiate breast collagenous spherulosis (positive) from adenoid cystic carcinoma. Adenoid cystic carcinoma in salivary gland tumors is calponin positive.

Host Species: Rabbit

Clone: EP798Y

Species Reactivity: Human. Others not known.

Immunogen: A synthetic peptide derived from near the C-terminus of human Calponin-1

Molecular Weight: 29/34 kDa

Positive Control: Breast CA

Cellular Localization: Cytoplasm Myoepithelial cells

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Calponin
Monoclonal
Unconjugated
A synthetic peptide derived from near the C-terminus of human Calponin-1
IVD
Primary
Immunohistochemistry (Paraffin)
EP798Y
Rabbit
12 mL
Cancer and Tumor Biology
Ready-to-Use
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.