missing translation for 'onlineSavingsMsg'
Learn More

CD35/CR1 (Follicular Dendritic Cell Marker) Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence and flow cytometry experiments.

Brand:  Epredia MS-336-P1

 View more versions of this product

Product Code. 12671037

  • ONLINE EXCLUSIVE PRICE
    435.00€
    363.83€ / 500µL
    Save 71.17€
    16% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Complement receptor 1 (CR1, CD35) is a cell membrane-bound, monomeric glycoprotein that is found on numerous cell types of primates including erythrocytes, lymphocytes, and phagocytes. The primary function of CR1 is to serve as the cellular receptor for C3b and C4b, the most important components of the complement system leading to clearance of foreign macromolecules.

Host Species: Mouse

Clone: E11

Isotype: IgG1

Species Reactivity: Human, Rhesus Monkey and Baboon. Others not tested.

Epitope: Distal from the C3b-binding site

Immunogen: Intact human monocytes

Molecular Weight: 210-220 kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD35
Monoclonal
800μg/mL
Mouse
500 μL
Hematopoietic
Human, Non-human Primate
IgG1
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
E11
Unconjugated
Intact human monocytes
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.