missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CD5 Ab-1, Mouse Monoclonal Antibody

Obtain accurate and reproducible results in immunohistochemistry applications with Epredia™ CD5 Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-393-RQ

 View more versions of this product

Product Code. 12651297

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

This item is not returnable. View return policy

Description

Description

CD5, a transmembrane protein, is found on 95% of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. CD5 is expressed by many T cell leukemia, lymphoma, and activated T cells. Occasionally, CD5 antigen is also expressed on a subset of B cells. Mantle cell lymphomas (same as diffuse centrocytic lymphomas) are CD5+ while the follicle center cell lymphoma are CD5-.

Host Species: Mouse

Clone: 4C7

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: External domain

Immunogen: Recombinant protein corresponding to the external domain of the CD5 molecule

Molecular Weight: 67kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD5 Ab-1
Monoclonal
Unconjugated
Recombinant protein corresponding to the external domain of the CD5 molecule
IVD
Primary
Immunohistochemistry (Paraffin)
4C7
Mouse
12 mL
Hematopoietic
Ready-to-Use
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.