missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CEA (Carcinoembryonic Antigen)/CD66e Ab-2 Rabbit Polyclonal Antibody with BSA and azide

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments.

102.00€

Specifications

Antigen CEA (Carcinoembryonic Antigen)/CD66e Ab-2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Product Code Brand Quantity Form Price Quantity & Availability  
Product Code Brand Quantity Form Price Quantity & Availability  
12623257
View Documents
Epredia™
RB-368-A0
100 μL Concentrated This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
Description

Description

Carcinoembryonic antigen (CEA), is synthesized during development in the fetal gut, and is re-expressed in increased amounts in intestinal carcinomas and several other tumors. Antibody to CEA is reportedly useful in identifying the origin of various metastatic adenocarcinomas and in distinguishing pulmonary adenocarcinomas (60-70% are CEA+) from pleural mesotheliomas (rarely or weakly CEA+).

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Mouse. Weakly reacts with Rat. Does not react with dog. Others not known

Immunogen: Human CEA isolated from hepatic metastasis of colon adenocarcinoma

Molecular Weight: 180 kDa

Positive Control: LS174T cells or colon CA

Cellular Localization: Cytoplasmic and luminal membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native and denatured)

Specifications

Specifications

CEA (Carcinoembryonic Antigen)/CD66e Ab-2
Polyclonal
Rabbit
Oncogenesis
Primary
Immunoprecipitation, Western Blot
Unconjugated
IVD
Human CEA isolated from hepatic metastasis of colon adenocarcinoma
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.