Learn More
Chromogranin A Ab-2 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Epredia™ Chromogranin A Ab-2, Mouse Monoclonal Antibody.
Brand: Epredia MS-381-PABX
Additional Details : Weight : 1.00000kg
Description
Chromogranin A (a protein of 439-amino acid which is encoded on chromosome 14) is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, pheo-chromocytomas, paragangliomas, and other neuro-endocrine tumors. Coexpression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms.Host Species: Mouse
Clone: PHE5
Isotype: IgG1
Species Reactivity: Human and Monkey. Does not react with rat. Others not known.
Immunogen: Human pheochromocytoma
Molecular Weight: 68-75 kDa
Positive Control: Pancreas
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifications
Chromogranin A Ab-2 | |
Monoclonal | |
1mg/mL | |
Mouse | |
200 μL | |
Oncogenesis | |
Human, Non-human Primate | |
IgG1 |
Immunofluorescence | |
PHE5 | |
Unconjugated | |
Human pheochromocytoma | |
RUO | |
Primary | |
Concentrated |