Learn More
Chromogranin A Rabbit Polyclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Chromogranin A, Rabbit Polyclonal Antibody.
Brand: Epredia RB-9003-P
Additional Details : Weight : 0.00250kg
Description
Chromogranin A (a protein of 439 amino acid which is encoded on chromosome 14) is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, phenochromocytomas, paragangliomas, and other neuroendocrine tumors. Coexpression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms. This antibody is excellent for formalin-fixed, paraffin-embedded tissues with no special pretreatment.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Monkey, Pig, Rat and Rabbit. Others not known
Epitope: Not determined
Immunogen: Recombinant protein encoding human chromogranin A
Molecular Weight: 68-75 kDa
Positive Control: Pancreas
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Chromogranin A | |
Polyclonal | |
200µg/mL | |
Rabbit | |
1 mL | |
Oncogenesis | |
Human, Non-human Primate, Porcine, Rabbit | |
IgG |
Immunohistochemistry (Paraffin) | |
Polyclonal | |
Unconjugated | |
Recombinant protein encoding human chromogranin A | |
RUO | |
Primary | |
Concentrated |