missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ COX2 Rabbit Monoclonal Antibody

Obtain accurate, reproducible results in Immunohistochemistry experiments with Epredia™ COX2, Rabbit Monoclonal Antibody, part of a comprehensive antibody portfolio.

Brand:  Epredia™ RM-9121-S1

Additional Details : Weight : 4.98950kg

 View more versions of this product

Product Code. 12613777

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.


Description

Description

COX2 (Cyclooxygenase-2) is an inducible enzyme. It is involved in the response of cells to growth factors, tumor promoters, and cytokines that induce its expression. Given its role in synthesizing prostaglandins, COX2 is therefore of interest in studying immune response regulation. COX2 is induced by a wide variety of stimuli and was initially identified as immediate-early growth response gene. In addition, COX2 expression markedly increased in 85-90% of human colorectal adenocarcinoma whereas COX1 levels remain unchanged.

Host Species: Rabbit

Clone: SP21

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: C-terminus

Immunogen: A synthetic peptide from C-terminus of rat COX2

Molecular Weight: 70 kDa

Positive Control: Lung and colon CAs

Cellular Localization: Cytoplasmic and membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

COX2
Monoclonal
Unconjugated
A synthetic peptide from C-terminus of rat cox2
IVD
Primary
Concentrated
Immunohistochemistry (Paraffin)
SP21
Rabbit
500 μL
Enzyme
Human, Mouse
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions