missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Epstein-Barr Virus/LMP Ab-1 Mouse Monoclonal Antibody

Product Code. 12679856 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Unit Size:
100µL
1mL
500µL
7mL
Product Code. Quantity Form unitSize
12679856 7 mL Prediluted 7mL
12699856 100 μL Concentrated 100µL
12609866 500 μL Concentrated 500µL
12689856 1 mL Concentrated 1mL
4 options
This item is not returnable. View return policy

Product Code. 12679856

Brand: Epredia™ MS1458R7

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Epstein-Barr Virus/LMP Ab-1, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: CS1 + CS2 + CS3 + CS4

Isotype: IgG1/κ

Species Reactivity: EBV-positive cells

Epitope: Located in the internal part of the membrane-associated viral LMP

Immunogen: EBV-encoded recombinant latent membrane protein

Molecular Weight: 60 kDa (LMP)

Positive Control: EBV-positive lymphoblastoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Epstein-Barr Virus/LMP Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CS1+CS2+CS3+CS4
Conjugate Unconjugated
Host Species Mouse
Immunogen EBV-encoded recombinant latent membrane protein
Quantity 7 mL
Regulatory Status ASR, IVD
Research Discipline Infectious Disease
Primary or Secondary Primary
Target Species Ebola virus
Form Prediluted
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.