missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Human Papilloma Virus (HPV) Ab-3 Mouse Monoclonal Antibody

Recommended for Immunohistochemistry (Formalin/paraffin), Epredia™ Human Papilloma Virus (HPV) Ab-3, Mouse Monoclonal Antibody is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Brand:  Epredia™ MS-1826-RQ

 View more versions of this product

Product Code. 12600227

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

This item is not returnable. View return policy

Description

Description

Human papilloma virus is non-enveloped double stranded circular DNA virus with icosahedral capsid.

Host Species: Mouse

Clone: K1H8

Isotype: IgG1/κ

Species Reactivity: Human Papilloma Virus type 6, 11, 16, 18, 31, 33, 42, 51, 52, 56 and 58.

Epitope: Nonconfirmational internal linear epitope of major capsid protein

Immunogen: Alkaline disrupted HPV type 1

Molecular Weight: 57kDa (major band)

Positive Control: Infected cells and tissue

Cellular Localization: Nuclei of infected cells

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Human Papilloma Virus (HPV) Ab-3
Monoclonal
Unconjugated
Alkaline disrupted HPV type 1
ASR, IVD
Primary
Immunohistochemistry (Paraffin)
K1H8
Mouse
12 mL
Infectious Disease
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.