Learn More
Description
Host Species: Mouse
Clone: UJ127
Isotype: IgG1/k
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Homogenous suspension of 16-week human fetal brain
Molecular Weight: 220-240kDa
Positive Control: IMR-5 cells Neuroblastoma
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Native and denatured)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | L1 Cell Adhesion Molecule Ab-1 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | UJ127 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Homogenous suspension of 16 week human fetal brain |
| Quantity | 200 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.