Learn More
Epredia™ Lab Vision™ MMP-13 (Collagenase-3) Ab-1 Mouse Monoclonal Antibody with BSA and azide
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: VIIIA2
Isotype: IgG1/κ
Species Reactivity: Human, Mouse and Rabbit. Others not known.
Epitope: Not determined
Immunogen: Human recombinant collagenase-3 protein
Molecular Weight: ∽60kDa (proform) and ∽48kDa (active form)
Positive Control: Conditioned, serum-free medium from (TPA-treated) human chondrosarcoma SW1353 cells; Placenta; Bladder, breast and ovarian CAs
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunoprecipitation (Native verified)
- Affinity Purification (Not verified)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | MMP-13 (Collagenase-3) Ab-1 |
| Applications | Affinity Purification, Immunohistochemistry (Paraffin), Immunoprecipitation |
| Classification | Monoclonal |
| Clone | VIIIA2 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Human recombinant collagenase-3 protein |
| Quantity | 500 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.