missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ p53 Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Achieve accurate, reproducible results with the Epredia™ p53 Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-104-P0

 View more versions of this product

Product Code. 12678706

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

This item is not returnable. View return policy

Description

Description

p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to mdm2, SV40 T antigen and human papilloma virus E6 protein p53 senses DNA damage and possibly facilitating repair. Mutation involving p53 is found in a wide variety of malignant tumors, including breast, ovarian, bladder, colon, lung, and melanoma.

Host Species: Mouse

Clone: PAb 240

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: aa 212-217

Immunogen: Murine p53 protein corresponding to aa 14-289

Molecular Weight: 53kDa

Positive Control: MDA-231 or SKBR-3 cells, Colon CA

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Methacarn or formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native and denatured)
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

p53 Ab-1
Monoclonal
200μg/mL
Mouse
100 μL
Cell Cycle
Human
IgG1
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
PAb 240
Unconjugated
Murine p53 protein corresponding to aa14-289
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.