Learn More
Pan-Cytokeratin Ab-2 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Epredia™ Cytokeratin, Pan Ab-2, Mouse Monoclonal Antibody.
Brand: Epredia MS-356-P1
Additional Details : Weight : 1.00000kg
Description
Twenty human keratins are resolved with two-dimensional gel electrophoresis into acidic (pI <5.7) and basic (pI >6.0) subfamilies. Members of the acidic and basic subfamilies are found together in pairs. The composition of keratin pairs varies with the epithelial cell type, stage of differentiation, cellular growth environment, and disease state. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.Host Species: Mouse
Clone: PAN-CK
Isotype: IgGs
Species Reactivity: Human. Others not known.
Epitope: Not determined
Positive Control: Skin
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifications
Cytokeratin, Pan Ab-2 | |
Monoclonal | |
200μg/mL | |
Mouse | |
RUO | |
Primary | |
Concentrated |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
PAN-CK | |
Unconjugated | |
500 μL | |
Cytoskeletal Protein | |
Human |